Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel QuickVue™ Strep A Swabs - 50/Pk
SDP

Catalog No. 23-038-171 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
50 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038-171 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-038-171 Supplier Quidel Supplier No. 20227
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with QuickVue Dipstick Strep A test and QuickVue+ Strep A kit only.

50 individually pouched sterile Rayon swabs.

Specifications

Detectable Analytes Streptococcal Antigen (Group A)
Sample Type Throat Swab
Type Streptococcus A Swab
Material Rayon
Packaging Type 50 Individually Pouched
Quantity 50 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.