Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quidel QuickVue™ iFOB Test

Click to view available options
Quantity:
1 Case
12 Case
6 Case
8 Case
Type:
iFOB Control Set
iFOB Spec Collect, Return Kit
iFOB Specimen Collection Kit
iFOB Test Kit
iFOBT Cassette Kit
Description
Includes
Individually wrapped test cassettes, package insert, procedure card, specimen collection and return kits, each consisting of: Specimen collection tube, with 2mL FOB buffer (1 x PBS with 0.02% sodium azide) and patient identification label, specimen collection paper with adhesive, specimen pouch, absorbent sleeve, return mailer, patient instructions
- CLIA Complexity: Waived
- Specificity: Specific to human hemoglobin
- Analytical Sensitivity: 50ng hHb/mL buffer or 50μg hHb/g feces
- POL agreement: 98%
- Results in 5 to 10 minutes; positive results may become visible sooner
- Patient friendly, easy to use
- Patients are not required to adhere to strict dietary restrictions
Order Info
- Additional specimen collection and return kits available separately in convenient 10-kit size
- External controls available separately
Specifications
Specifications
Clia Complexity | Waived |
Type | iFOBT Cassette Kit |
For Use With (Application) | To detect the presence of Blood in stool specimens |
Includes | Individually wrapped test cassettes, package insert, procedure card, specimen collection and return kits, each consisting of specimen collection tube, with 2mL FOB Buffer (1 x PBS with 0.02% Sodium Azide) and patient identification label, specimen collection Paper with adhesive, specimen pouch, absorbent sleeve, return mailer, patient instructions |
Quantity | 6 Case |
Test Time | 5 to 10 min. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction