Learn More
Description
Includes
Individually wrapped test cassettes, package insert, procedure card, specimen collection and return kits, each consisting of: Specimen collection tube, with 2mL FOB buffer (1 x PBS with 0.02% sodium azide) and patient identification label, specimen collection paper with adhesive, specimen pouch, absorbent sleeve, return mailer, patient instructions
- CLIA Complexity: Waived
- Specificity: Specific to human hemoglobin
- Analytical Sensitivity: 50ng hHb/mL buffer or 50μg hHb/g feces
- POL agreement: 98%
- Results in 5 to 10 minutes; positive results may become visible sooner
- Patient friendly, easy to use
- Patients are not required to adhere to strict dietary restrictions
Order Info
- Additional specimen collection and return kits available separately in convenient 10-kit size
- External controls available separately
Specifications
Specifications
| Clia Complexity | Waived |
| Type | iFOB Test Kit |
| For Use With (Application) | To detect the presence of Blood in stool specimens |
| Includes | Individually wrapped test cassettes, package insert, procedure card, specimen collection and return kits, each consisting of specimen collection tube, with 2mL FOB Buffer (1 x PBS with 0.02% Sodium Azide) and patient identification label, specimen collection Paper with adhesive, specimen pouch, absorbent sleeve, return mailer, patient instructions |
| Quantity | 6 Case |
| Test Time | 5 to 10 min. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
