Learn More
Quidel Sofia™ hCG FIA
SDP
Catalog No. 23043078
Click to view available options
Quantity:
3 x 50 Tests
50 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy
Kinetic Check™ feature provides operator the flexibility of upto 5 minutes before placing test cassette into Sofia for analysis
  • Specific to monoclonal antibodies of beta subunit of hCG
  • Eliminates operator to operator variability and reduces transcription and billing errors
  • Automated results in three minutes
  • High accuracy and consistency
  • Prints results and/or sends to LIS for easy transfer of patient records
  • 20mIU/mL sensitivity in urine
  • Store at room temperature, 18 months shelf life from date of manufacture
Order Info
Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information.
Specifications
Type hCG Starter Kit
Quantity 3 x 50 Tests
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.