Learn More
Quidel Sofia™ hCG FIA
SDP
Catalog No. p-4706593 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
23-043-077 50 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-043-077 Supplier Quidel Supplier No. 20229
This item is not returnable. View return policy
Kinetic Check™ feature provides operator the flexibility of upto 5 minutes before placing test cassette into Sofia for analysis
  • Specific to monoclonal antibodies of beta subunit of hCG
  • Eliminates operator to operator variability and reduces transcription and billing errors
  • Automated results in three minutes
  • High accuracy and consistency
  • Prints results and/or sends to LIS for easy transfer of patient records
  • 20mIU/mL sensitivity in urine
  • Store at room temperature, 18 months shelf life from date of manufacture
Specifications
Type hCG FIA Starter Kit
Quantity 50 Tests