Learn More
Contec™ Quiltec™ Mop Head
BIOPHARMA
Catalog No. 19168582 Shop All Contec Inc. Products
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
19-168-582 Non-sterile
1 options
Catalog No. 19-168-582 Supplier Contec™ Supplier No. GCQT0100
Only null left
Add to Cart
Add to Cart
Manufactured from 2-ply polyester around a rayon core. Contec™ Quiltec Mop Head is ideal for the highest grade of cleanroom due to the small numbers of particle and fibers.
  • Two-ply 100% knitted polyester over a rayon core
  • Gamma irradiated at no less than 25 kGy
  • 5 mops double bagged
  • Approx 18 to 20 sqm coverage
  • Certificates of conformity supplied with every batch
Specifications
Color Code White
Product Type Mop Head
Sterility Non-sterile
Autoclavable Not Autoclavable
Disposable Single-use
Quantity 40/Cs.
Description Quiltec Polyester Mop Head
Color-coded No
Material Quilted Polyester