Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinaldic acid, 98%

Catalog No. AAA1019214
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AAA1019214

Supplier: Thermo Scientific Chemicals A1019214

Only null left
Add to Cart

CAS: 93-10-7 | C10H7NO2 | 173.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-10-7
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00006752
InChI Key LOAUVZALPPNFOQ-UHFFFAOYSA-N
Synonym quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid
PubChem CID 7124
ChEBI CHEBI:18386
IUPAC Name quinoline-2-carboxylic acid
SMILES OC(=O)C1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 156°C to 159°C
Quantity 25 g
Beilstein 126322
Merck Index 14,8046
Formula Weight 173.17
Percent Purity 98%
Chemical Name or Material Quinaldic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-218-3
RTECSNumber UZ9100000
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.