Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quinaldine, ≥90% (GC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
500 mL
1 L
Description
Pharmaceutical (small molecule)

Specifications
Specifications
Packaging | Glass Bottle |
CAS | 91-63-4 |
Density | 1.058 g/mL (at 25°C) |
Flash Point | 79°C |
Physical Form | Neat |
Linear Formula | C10H9 N |
MDL Number | MFCD00006756 |
Melting Point | -9°C to -3°C |
Boiling Point | 105°C to 107°C (10 mmHg); 248°C |
Synonym | 2-Methylquinoline |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction