Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinazoline, 98%

Catalog No. AC153980050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

CAS: 253-82-7 | C8H6N2 | 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253-82-7
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006712
InChI Key JWVCLYRUEFBMGU-UHFFFAOYSA-N
Synonym phenmiazine, 1,3-benzodiazine, 1,3-diazanaphthalene, 5,6-benzopyrimidine, benzo a pyrimidine, chinazolin, unii-ub9qur18nl, ub9qur18nl, benzopyrimidine
PubChem CID 9210
ChEBI CHEBI:36621
IUPAC Name quinazoline
SMILES C1=CC=C2N=CN=CC2=C1

Specifications

Melting Point 45°C to 48°C
Color Brown to Yellow
Boiling Point 243°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 23, 175
Merck Index 15, 8169
Formula Weight 130.15
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Quinazoline, 98%
Show More Show Less
EINECSNumber 205-965-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.