Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Quinidine Monoclonal Antibody (QD11-261.3.6)
GREENER_CHOICE

Catalog No. 01671977
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-977 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-977 Supplier Invitrogen™ Supplier No. MA110747
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10747 detects Quinidine. MA1-10747 has been successfully used in ELISA applications. The MA1-10747 immunogen is Quinidine, conjugated to KLH.

Quinidine is an alkaloid extracted from the bark of the Cinchona tree with class 1A antiarrhythmic and antimalarial effects. Quinidine stabilizes the neuronal membrane by binding to and inhibiting voltage-gated sodium channels, thereby inhibiting the sodium influx required for the initiation and conduction of impulses resulting in an increase of the threshold for excitation and decreased depolarization during phase 0 of the action potential. In addition, the effective refractory period (ERP), action potential duration (APD), and ERP/APD ratios are increased, resulting in decreased conduction velocity of nerve impulses. Quinidine exerts its antimalarial activity by acting primarily as an intra-erythrocytic schizonticide through association with the heme polymer (hemazoin) in the acidic food vacuole of the parasite thereby preventing further polymerization by heme polymerase enzyme. This results in accumulation of toxic heme and death of the parasite. [National Center for Biotechnology Information. PubChem Compound Database; CID=441074].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Quinidine
Applications ELISA
Classification Monoclonal
Clone QD11-261.3.6
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C20H24N2O2; Chinidin; Cin-Quin; Pitayine; Quinidex
Host Species Mouse
Immunogen Quinidine, conjugated to KLH.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.