Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Quinine Sulfate, Dihydrate
SCI_ED

Catalog No. p-4416538 Shop All Fisherbrand Products
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Lab Grade - Powder

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 6119-70-6
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
MDL Number MFCD00150790
InChI Key ZHNFLHYOFXQIOW-OIGVVMIYNA-N
Synonym Quinine Sulfate, Quinine Bisulfate
PubChem CID 134129495
IUPAC Name bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
SMILES O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Color White
Assay Percent Range >99%
Packaging Lab Grade - Powder
Linear Formula (C20H24N2O2)2·H2SO4·2H2O
Quantity 5 g
Formula Weight 782.97
Grade Laboratory
Chemical Name or Material Quinine Sulfate, Dihydrate
ShelfLife 5 years
DOTInformation DOT Class 6.1, III, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.