Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinine sulfate dihydrate, specified according to the requirements of USP

Catalog No. ac418780050
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle

CAS: 6119-70-6 | C40H54N4O10S | 782.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6119-70-6
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
MDL Number MFCD00150790
InChI Key ZHNFLHYOFXQIOW-OIGVVMIYNA-N
PubChem CID 134129495
SMILES O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Melting Point 153.0°C
Ignition Residue 0.1% max
Packaging Glass Bottle
Quantity 5 g
Merck Index 15,8177
IUPAC Name bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Formula Weight 782.96
Insoluble Matter 0.1% max. (in mixture of chloroform-alcohol (2:1))
Chemical Name or Material Quinine Sulfate Dihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.