Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinine Sulfate, Dihydrate, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606023
Click to view available options
Quantity:
10 g
100 g
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

QU110, 207671-44-1, (C20H24N2O2)2·H2SO4·2H2O

Spectrum™ Chemical Quinine Sulfate, Dihydrate, USP is a medication used to treat malaria and babesiosis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 207671-44-1

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 10 g
Specific Rotation -235° to -245°
Percent Purity 99 to 101%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.