Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoline, 96%

Catalog No. AC132195000
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

CAS: 91-22-5 | C9H7N | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-22-5
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987
InChI Key SMWDFEZZVXVKRB-UHFFFAOYSA-N
Synonym chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol
PubChem CID 7047
ChEBI CHEBI:17362
IUPAC Name quinoline
SMILES C1=CC=C2N=CC=CC2=C1

Specifications

Melting Point -15.0°C
Color Brown to Yellow
Density 1.0950g/mL
Boiling Point 237.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.6230 to 1.6260
Quantity 500 mL
Beilstein 20,339
Fieser 01,975; 02,356; 09,403
Merck Index 15,8185
Specific Gravity 1.095
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in hot water,miscible with alcohol,ether,acetone,benzene,and carbon disulfide
Formula Weight 129.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Quinoline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful in contact with skin.
Toxic
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Call a POISON CENT
EINECSNumber 202-051-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.