Learn More
Quinoline, 96%
Description
Quinoline is used as a chemical intermediate for making medicines and other organic compounds. It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Quinoline is used as a chemical intermediate for making medicines and other organic compounds. It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.
Solubility
More soluble in hot than cold water. Miscible with alcohol, ether
Notes
Dissolves sulfur, As{2}O{3}, phosphorus

Specifications
Specifications
Melting Point | -15°C |
Density | 1.09 |
pH | 7.3 |
Boiling Point | 237°C |
Flash Point | 101°C (214°F) |
Refractive Index | 1.626 |
Quantity | 2500 g |
UN Number | UN2656 |
Beilstein | 107477 |
Sensitivity | Hygroscopic; Light sensitive |
Show More |
Safety and Handling
Hazard Category | H301-H312-H315-H319-H341-H350 |
Hazard Statement | GHS H Statement H311-H350-H341-H302-H315-H319 Toxic in contact with skin. May cause cancer. Suspected of causing genetic defects. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P201-P202-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: QUINOLINE |
EINECSNumber | 202-051-6 |
RTECSNumber | VA9275000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.