Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quinoline, 96%
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Quinoline is used as a chemical intermediate for making medicines and other organic compounds. It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.
Solubility
More soluble in hot than cold water. Miscible with alcohol, ether
Notes
Dissolves sulfur, As{2}O{3}, phosphorus
Quinoline is used as a chemical intermediate for making medicines and other organic compounds. It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.
Solubility
More soluble in hot than cold water. Miscible with alcohol, ether
Notes
Dissolves sulfur, As{2}O{3}, phosphorus
Specifications
Specifications
| Melting Point | -15°C |
| Density | 1.09 |
| pH | 7.3 |
| Boiling Point | 237°C |
| Flash Point | 101°C (214°F) |
| Refractive Index | 1.626 |
| Quantity | 100 g |
| UN Number | UN2656 |
| Beilstein | 107477 |
| Sensitivity | Hygroscopic; Light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H301-H312-H315-H319-H341-H350 |
| Hazard Statement | GHS H Statement H311-H350-H341-H302-H315-H319 Toxic in contact with skin. May cause cancer. Suspected of causing genetic defects. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P201-P202-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: QUINOLINE |
| EINECSNumber | 202-051-6 |
| RTECSNumber | VA9275000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction