Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinolinic acid, Tocris Bioscience™

Catalog No. 02251g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Endogenous NMDA agonist and transmitter candidate

Endogenous NMDA agonist and transmitter candidate. May distinguish between NMDA receptor subtypes.

Chemical Identifiers

CAS 89-00-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
InChI Key GJAWHXHKYYXBSV-UHFFFAOYSA-N
Synonym quinolinic acid, 2,3-pyridinedicarboxylic acid, quinolinate, pyridine-2,3-dicarboxylate, unii-f6f0hk1urn, 2,3-pyridinedicarboxylicacid, pyridine-2,3-carboxylate, f6f0hk1urn, pyridin-2,3-dicarbonsaeure, 2,3-pyridinedicarboxylate
PubChem CID 1066
ChEBI CHEBI:16675
IUPAC Name pyridine-2,3-dicarboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)C(=O)O

Specifications

Quantity 1 g
Formula Weight 167.12
Percent Purity >98%
Chemical Name or Material Quinolinic acid
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.