Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) acetate, 99.99%, (trace metal basis), anhydrous

Catalog No. AC436980500
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

CAS: 25114-58-3 | C6H9InO6 | 291.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25114-58-3
Molecular Formula C6H9InO6
Molecular Weight (g/mol) 291.95
InChI Key LGHOYKSQIQISBI-UHFFFAOYSA-N
Synonym indium acetate, acetic acid, indium 3+ salt
PubChem CID 24191761
IUPAC Name acetic acid;indium
SMILES CC(=O)O.[In]

Specifications

Melting Point 270.0°C
Color White
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 35.3 to 43.3% (In) (Complexometry)
Solubility Information Solubility in water: soluble
Formula Weight 291.95
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Indium(III) acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.