Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QX 222, Tocris Bioscience™

Catalog No. p-4785408
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Na+ channel blocker

Sodium channel blocker.

Chemical Identifiers

CAS 5369-00-6
Molecular Formula C13H21ClN2O
Molecular Weight (g/mol) 256.774
InChI Key WFKXSWWTOZBDME-UHFFFAOYSA-N
Synonym lidocaine n-methyl hydrochloride, 2,6-dimethylphenyl carbamoyl methyl trimethylazanium chloride
PubChem CID 9795082
IUPAC Name [2-(2,6-dimethylanilino)-2-oxoethyl]-trimethylazanium;chloride
SMILES CC1=C(C(=CC=C1)C)NC(=O)C[N+](C)(C)C.[Cl-]

Specifications

Quantity 10 mg
Formula Weight 256.77
Percent Purity >98%
Chemical Name or Material QX 222
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.