Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ QX 314 bromide
GREENER_CHOICESDP

Catalog No. 1014100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-141-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-141-00 Supplier Tocris Bioscience™ Supplier No. 1014/100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Na+ channel blocker

Membrane impermeable quaternary derivative of lidocaine, a blocker of voltage-activated Na+ channels. Intracellular QX 314 bromide also inhibits calcium currents in hippocampal CA1 pyramidal neurons.

Chemical Identifiers

CAS 24003-58-5
Molecular Formula C16H27BrN2O

Specifications

Quantity 100 mg
Formula Weight 343.31
Purity >99%
Molecular Weight (g/mol) 343.309
Chemical Name or Material QX 314 bromide
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.