Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ QX 314 chloride
GREENER_CHOICESDP

Catalog No. 231350 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-135-0 Supplier R&D Systems™ Supplier No. 2313/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Na+ channel blocker

Membrane impermeable quaternary derivative of lidocaine, a blocker of voltage-activated Na+ channels.

Chemical Identifiers

CAS 1-4-5370
Molecular Formula C11H18ClNO

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 298.85
Purity >99%
Molecular Weight (g/mol) 215.72
Chemical Name or Material QX 314 chloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.