Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonyl cyanide 3-chlorophenylhydrazone, 98%

Catalog No. AC228131000
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 555-60-2 | C9H5ClN4 | 204.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-60-2
Molecular Formula C9H5ClN4
Molecular Weight (g/mol) 204.62
MDL Number MFCD00001848
InChI Key UGTJLJZQQFGTJD-UHFFFAOYSA-N
Synonym cccp, 3-chlorophenyl hydrazonomalononitrile, carbonylcyanide-3-chlorophenylhydrazone, carbonyl cyanide 3-chlorophenylhydrazone, carbonyl cyanide m-chlorophenyl hydrazone, carbonyl cyanide m-chlorophenylhydrazone, 3-chlorophenyl hydrazono malononitrile, m-chlorophenyl carbonylcyanide hydrazone, m-cl-ccp, carbonyl cyanide, 3-chlorophenyl hydrazone
PubChem CID 2603
ChEBI CHEBI:3259
IUPAC Name 2-[(3-chlorophenyl)hydrazinylidene]propanedinitrile
SMILES C1=CC(=CC(=C1)Cl)NN=C(C#N)C#N

Specifications

Melting Point 172°C to 177°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula ClC6H4NHN=C(CN)2
Quantity 100 mg
Solubility Information Solubility in water: insoluble
Formula Weight 204.62
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Carbonyl cyanide 3-chlorophenylhydrazone, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 209-103-7
RTECSNumber FG5600000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.