Learn More
(R)-(-)-1,3-Butanediol, 98%
Description
(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Specifications
Specifications
| Density | 1.005 |
| Boiling Point | 203°C to 204°C |
| Flash Point | 121°C (249°F) |
| Odor | Weak |
| Molecular Formula | C8H15NO4 |
| Refractive Index | 1.44 |
| Quantity | 1 g |
| Beilstein | 1718944 |
| Sensitivity | Hygroscopic |
| Synonym | diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799 |
| Show More |
Safety and Handling
| EINECSNumber | 228-532-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.