Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1-Amino-2-propanol 98.0+%, TCI America™
SDP

Catalog No. A097425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A097425G 25 g
A09741G 1 g
A09745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A097425G Supplier TCI America Supplier No. A097425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2799-16-8
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064428
InChI Key HXKKHQJGJAFBHI-GSVOUGTGSA-N
Synonym r---1-amino-2-propanol, r-1-aminopropan-2-ol, r-1-amino-2-propanol, 2r-1-aminopropan-2-ol, r---isopropanolamine, 2-propanol, 1-amino-, 2r, 2r---hydroxypropylamine, r---1-aminopropan-2-ol, 2r---2-hydroxypropylamine
PubChem CID 439938
ChEBI CHEBI:15675
IUPAC Name (2R)-1-aminopropan-2-ol
SMILES C[C@@H](O)CN

Specifications

Color White-Yellow
Boiling Point 160°C
Quantity 25 g
UN Number 3259
Formula Weight 75.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (R)-(-)-1-Amino-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.