Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-1-Benzyl-3-hydroxypiperidine 98.0+%, TCI America™
SDP

Catalog No. B48575G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 91599-81-4
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00274307
InChI Key UTTCOAGPVHRUFO-GFCCVEGCSA-N
Synonym r-1-benzylpiperidin-3-ol, r-1-benzyl-3-hydroxypiperidine, 3r-1-benzylpiperidin-3-ol, r-1-n-benzyl-3-hydroxy-piperidine, r-1-benzyl-3-piperidinol, r---1-benzyl-3-hydroxypiperidine, r-n-benzyl-3-hydroxypiperidine, 3-piperidinol, 1-phenylmethyl-, 3r, r-pound inverted question mark-1-benzyl-3-hydroxypiperidinol, pubchem13101
PubChem CID 693761
IUPAC Name (3R)-1-benzylpiperidin-3-ol
SMILES C1CC(CN(C1)CC2=CC=CC=C2)O

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
Formula Weight 191.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-1-Benzyl-3-hydroxypiperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.