Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-1-Benzyl-3-pyrrolidinecarbonitrile 98.0+%, TCI America™
SDP

Catalog No. B32991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32991G 1 g
B32995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32991G Supplier TCI America Supplier No. B32991G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 157528-56-8
Molecular Formula C12H14N2
Molecular Weight (g/mol) 186.258
MDL Number MFCD09951967
InChI Key RYCQUUNQHVAFSM-LBPRGKRZSA-N
Synonym (R)-1-Benzyl-3-cyanopyrrolidine
PubChem CID 1515497
IUPAC Name (3R)-1-benzylpyrrolidine-3-carbonitrile
SMILES C1CN(CC1C#N)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 186.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-1-Benzyl-3-pyrrolidinecarbonitrile
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.