Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1-Methoxy-2-propanol 98.0+%, TCI America™
SDP

Catalog No. M216625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M216625G 25 g
M21665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M216625G Supplier TCI America Supplier No. M216625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4984-22-9
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD01632587
InChI Key ARXJGSRGQADJSQ-SCSAIBSYSA-N
Synonym r---1-methoxy-2-propanol, 2r-1-methoxypropan-2-ol, r-1-methoxypropan-2-ol, r-1-methoxy-2-propanol, r-1-methoxy-propan-2-ol, 2-propanol, 1-methoxy-, 2r, 20s-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxahenicosane-1,20-diol, pubchem6714, r-methoxy-propan-2-ol
PubChem CID 2733589
IUPAC Name (2R)-1-methoxypropan-2-ol
SMILES CC(COC)O

Specifications

Color Colorless
Boiling Point 120°C
Quantity 25 g
UN Number 3092
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-Methoxy-2-propanol
EINECSNumber (2)-0404
RTECSNumber UB7700000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.