Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-Phenylpropylamine, ChiPros 99+%, ee 98%

Catalog No. p-7046780
Click to view available options
Quantity:
5 g
25 g

CAS: 3082-64-2 | C9H13N | 135.21 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3082-64-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00083057
InChI Key AQFLVLHRZFLDDV-SECBINFHSA-N
Synonym r-+-1-phenylpropylamine, r-1-phenylpropan-1-amine, 1r-1-phenylpropan-1-amine, r-1-phenylpropylamine, r-+-alpha-ethylbenzylamine, 1r-1-phenylpropylamine, r-+-methylbenzylamine, r-+-1-amino-1-phenylpropane, r +-a-ethylbenzylamine, r-+-, a-ethylbenzylamine
PubChem CID 5324978
IUPAC Name (1R)-1-phenylpropan-1-amine
SMILES CCC(C1=CC=CC=C1)N

Specifications

Melting Point -69°C
Density 0.93
Boiling Point 205°C
Flash Point 77°C (170°F)
Odor Amine-like
Refractive Index 1.52
Quantity 5 g
UN Number UN2735
Beilstein 3029904
Sensitivity Air Sensitive
Optical Rotation +20° (Neat)
Formula Weight 135.21
Percent Purity ≥99%
Assay ee 98%
Chemical Name or Material (R)-(+)-1-Phenylpropylamine
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 608-544-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.