Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%

Catalog No. p-7027208
Click to view available options
Quantity:
1 g

Catalog No. AAB2378503

Supplier: Thermo Scientific Chemicals B2378503

Only null left
Add to Cart

CAS: 76189-55-4 | C44H32P2 | 622.69 g/mol

(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthy is used as a chiral ligand for metal mediated asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthy is used as a chiral ligand for metal mediated asymmetric catalysis.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76189-55-4
Molecular Formula C44H32P2
Molecular Weight (g/mol) 622.69
MDL Number MFCD00010805
InChI Key MUALRAIOVNYAIW-UHFFFAOYSA-N
Synonym binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, s-binap, s---binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, 2,2'-bis diphenylphosphino-1,1'-binaphthalene
PubChem CID 634876
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 239°C to 241°C
Quantity 1 g
Beilstein 4914063
Sensitivity Air Sensitive
Merck Index 14,1223
Solubility Information Insoluble in water.
Optical Rotation +230° (c=0.3 in Toluene)
IUPAC Name [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
Formula Weight 622.69
Percent Purity 98%
Chemical Name or Material (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.