Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2,2'-Diamino-1,1'-binaphthalene, 99%, Thermo Scientific Chemicals

Catalog No. AC356510010
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18741-85-0
Molecular Formula C20H16N2
Molecular Weight (g/mol) 284.36
MDL Number MFCD00145204
InChI Key DDAPSNKEOHDLKB-UHFFFAOYSA-N
Synonym 1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
PubChem CID 20571
IUPAC Name 1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
SMILES NC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 242.0°C to 244.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation 156
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 284.36
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (R)-(+)-2, 2'-Diamino-1, 1'-binaphthalene
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
missing translation for 'healthHazard3' GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.