Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2-Acetoxysuccinic anhydride, 98%, Thermo Scientific™

Catalog No. AA4436306
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79814-40-7
Molecular Formula C6H6O5
Molecular Weight (g/mol) 158.109
MDL Number MFCD00145214
InChI Key SSWJHSASZZAIAU-SCSAIBSYSA-N
Synonym r-+-2-acetoxysuccinic anhydride, r-2-o-acetylmalic anhydride, r-2,5-dioxotetrahydrofuran-3-yl acetate, +-o-acetyl-d-malic anhydride, 2,5-furandione, 3-acetyloxy dihydro-, r, o-acetyl d-malic anhydride, s-acetoxysuccinic anhydride, o-acetyl-d-malic anhydride, ksc496o4p, r-acetoxysuccinic anhydride
PubChem CID 9815264
IUPAC Name [(3R)-2,5-dioxooxolan-3-yl] acetate
SMILES CC(=O)OC1CC(=O)OC1=O

Specifications

Melting Point 56°C to 58°C
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Reacts in water.
Formula Weight 158.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)- (+)-2-Acetoxysuccinic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.