Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Amino-1-butanol, 98%

Catalog No. p-3777216
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle

Catalog No. AC244030250

Supplier: Thermo Scientific Chemicals 244030250

Only null left
Add to Cart

CAS: 5856-63-3 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5856-63-3
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00064419
InChI Key JCBPETKZIGVZRE-UHFFFAOYNA-N
Synonym r-2-aminobutan-1-ol, r---2-amino-1-butanol, 2r-2-aminobutan-1-ol, r-2-amino-1-butanol, 1-butanol, 2-amino-, 2r, unii-4f725v55ln, 2-amino-1-butanol, r, 2-amino-1-butanol,-, l---2-amino-1-butanol, 1-butanol, 2-amino-, r
PubChem CID 2723856
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(N)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -2°C
Color White
Density 0.9470g/mL
Boiling Point 172°C to 174°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4495 to 1.4535
Linear Formula C2H5CH(NH2)CH2OH
Quantity 25 mL
Beilstein 04,III,771
Specific Gravity 0.947
Merck Index 10, 429
Specific Rotation Condition −10° (19°C c=neat)
Specific Rotation -10
Solubility Information Solubility in water: miscible. Other solubilities: very soluble in most of the usual solvents,hardly soluble in aliphatic hydrocarbons
Formula Weight 89.14
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material (R)-(-)-2-Amino-1-butanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 227-476-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.