Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Amino-1-propanol, 98%, Thermo Scientific Chemicals

Catalog No. AC400440010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC400440010

Supplier: Thermo Scientific Chemicals 400440010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35320-23-1
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064413
InChI Key BKMMTJMQCTUHRP-GSVOUGTGSA-N
Synonym d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol
PubChem CID 80308
IUPAC Name (2R)-2-aminopropan-1-ol
SMILES CC(CO)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9650g/mL
Boiling Point 173°C to 176°C
Flash Point 83°C
Enantiomeric Excess 99% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4480 to 1.451
Linear Formula CH3CH(NH2)CH2OH
Quantity 1 g
Beilstein 04, III, 735
Specific Gravity 0.965
Specific Rotation -17
Solubility Information Solubility in water: soluble in water
Formula Weight 75.11
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material (R)-(-)-2-Amino-1-propanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.