Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2-Amino-3-benzyloxy-1-propanol, 97%, Thermo Scientific™

Catalog No. AC399620010
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 58577-87-0
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.23
InChI Key ZJUOMDNENVWMPL-SNVBAGLBSA-N
Synonym r-2-amino-3-benzyloxy propan-1-ol, h-serinol bzl, r-+-2-amino-3-benzyloxy-1-propanol, 2r-2-amino-3-benzyloxy propan-1-ol, 2-amino-3-benzyloxy-1-propanol, 1-propanol, 2-amino-3-phenylmethoxy-, 2r, r-2-amino-3-benzyloxy-1-propanol, serinol benzyl ether, ambotzhal1055, r-2-amino-3-benzyloxypropan-1-ol
PubChem CID 14118408
IUPAC Name (2R)-2-amino-3-phenylmethoxypropan-1-ol
SMILES C1=CC=C(C=C1)COCC(CO)N

Specifications

Melting Point 34°C to 37°C
Boiling Point 307°C
Flash Point 113°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Linear Formula C6H5CH2OCH2CH(NH2)CH2OH
Quantity 1 g
Formula Weight 181.23
Percent Purity 97%
Chemical Name or Material (R)-(+)-2-Amino-3-benzyloxy-1-propanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.