Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Amino-3-methyl-1-butanol, 98%

Catalog No. AC422470250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422470250 25 g Glass bottle
AC422470050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422470250 Supplier Thermo Scientific Chemicals Supplier No. 422470250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

CAS: 4276-09-9 | C5H13NO | 103.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4276-09-9
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00064297
InChI Key NWYYWIJOWOLJNR-YFKPBYRVSA-N
IUPAC Name (2R)-2-amino-3-methylbutan-1-ol
SMILES CC(C)[C@@H](N)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 32.0°C to 36.0°C
Color White to Yellow
Density 0.9310g/mL
Boiling Point 189.0°C to 190.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 0.931
Specific Rotation Condition -16.50 (20.00°C c = 10,EtOH)
Synonym 4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl
Specific Rotation -16.50
PubChem CID 77973
Formula Weight 103.16
Percent Purity 98%
Physical Form Low Melting Crystals or Crystalline Mass
Chemical Name or Material (R)-(-)-2-Amino-3-methyl-1-butanol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
Recommended Storage Refrigerator +4°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.