Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2-(BOC-Amino)-1,4-butanediol, 97%, Thermo Scientific Chemicals

Catalog No. ac435570010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC435570010 1 g
AC435570050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435570010 Supplier Thermo Scientific Chemicals Supplier No. 435570010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 397246-14-9
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
InChI Key KLRRFBSWOIUAHZ-SSDOTTSWSA-N
Synonym r-+-2-boc-amino-1,4-butanediol, r-tert-butyl 1,4-dihydroxybutan-2-yl carbamate, r-2-boc-amino-butane-1,4-diol, r-2-boc-amino-1,4-butanediol, tert-butyl n-2r-1,4-dihydroxybutan-2-yl carbamate, pubchem15259, ksc498c9f, 1r-3-hydroxy-1-hydroxymethyl propyl carbamic acid tert-butyl ester, r-2-boc-amino-butan-1,4-diol, 2r-tert-butoxycarbonylamino butane-1,4-diol
PubChem CID 11954482
IUPAC Name tert-butyl N-[(2R)-1,4-dihydroxybutan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CCO)CO

Specifications

Melting Point 66.4°C to 72.1°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Specific Rotation 7.8
Quantity 1 g
Formula Weight 205.25
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-2-(BOC-Amino)-1, 4-butanediol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.