Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(R)-(-)-2-Chloromandelic acid, 99%, Thermo Scientific™
Click to view available options
1g; Glass bottle
Specifications
CAS | 52950-18-2 |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Assay Percent Range | 99% |
Molecular Formula | C8H7ClO3 |
MDL Number | MFCD00798429 |
Synonym | r---2-chloromandelic acid, r-2-2-chlorophenyl-2-hydroxyacetic acid, r-2-chloromandelic acid, r-o-chloromandelic acid, 2-chloro-d-mandelic acid, 2r-2-2-chlorophenyl-2-hydroxyacetic acid, r-2-chlorophenyl hydroxy acetic acid, pubchem5742, ksc914s2l, r---2-chloromandelicacid |
InChI Key | RWOLDZZTBNYTMS-SSDOTTSWSA-N |
SMILES | C1=CC=C(C(=C1)C(C(=O)O)O)Cl |
IUPAC Name | (2R)-2-(2-chlorophenyl)-2-hydroxyacetic acid |
Show More |
Safety and Handling
Hazard Statement | GHS H Statement: Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
TSCA | TSCA |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction