Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (R)-2-hydroxy-4-methylpentanoic acid

Catalog No. CDS006369 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS006369 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS006369 Supplier Sigma Aldrich Supplier No. CDS00636950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-acetoxy-1,3-butadiene (1-abd) can be generated by potassium or sodium acetate catalyzed reaction between crotonaldehyde and acetic anhydride. the product is a mixture of cis and trans forms, that has been confirmed by its physical properties and IR spectra. it is reported to be formed as a major product during the acetoxylation of 1,3-butadiene (bd) in gas-phase in the presence of pd-koac (palladium-potassium acetate) catalyst. 1-abd participates as 2+ß or 4+ß diene in cycloaddition reactions. enantioselective diels alder reaction of 2-methoxy-5-methyl-1,4-benzoquinone with 1-abd in the presence of molecular sieves (mesh size:4+a) has been reported.1-acetoxy-1,3-butadiene was used as diene in the following reactions:
  • Diels-alder reaction with ortho-carbazolequinones to yield benzocarbazolequinone
  • Diels-alder reaction with diethyl ketovinylphosphonate, with and without lewis acid assistance
  • Diels-alder reaction with methyl acrylate to yield racemic forms of 2-hydroxy-3-cyclohexenecarboxylic acid. it was used for the reaction with dienophiles such as maleimides, a juglone, a butyne-1,4-dione and methyl 2-(4-methylphenyl)-2h-azirine-3-carboxylate and during visible light photocatalysis. it was also used as reactant during intermolecular oxa-pictet-spengler cyclization

Specifications

CAS 20312-37-2
Molecular Formula C6H12O3
Linear Formula C6H12O3
MDL Number MFCD00211263
Quantity 10 g
Molecular Weight (g/mol) 132.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.