Learn More
Tocris Bioscience™ (R)-2-Hydroxyglutaric acid disodium salt, Tocris Bioscience™

Shop All Novus Biologicals Products


Description
Specifications
Specifications
| Chemical Name or Material | (R)-2-Hydroxyglutaric acid disodium salt |
| CAS | 103404-90-6 |
| Synonym | (R)-2-Hydroxypentanedioic acid disodium salt |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in water |
| Molecular Formula | C5H6Na2O5 |
| Formula Weight | 192.08 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.