Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Phenylbutyric acid, 99%

Catalog No. AC378520050
Click to view available options
Quantity:
1 g
5 g

CAS: 938-79-4 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-79-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00063165
InChI Key OFJWFSNDPCAWDK-UHFFFAOYNA-N
Synonym r---2-phenylbutyric acid, 2r-2-phenylbutanoic acid, 2-phenylbutyric acid, r, r-2-phenylbutyric acid, benzeneacetic acid, alpha-ethyl-, r, r-2-phenylbutanoic acid, 2r-2-phenyl-butanoic acid, 2r-2-phenyl-mutanoic acid, r---2-phenylbutanoic acid, benzeneacetic acid,a-ethyl-, ar
PubChem CID 785330
IUPAC Name (2R)-2-phenylbutanoic acid
SMILES CCC(C(O)=O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.0550g/mL
Boiling Point 130°C (3.0 mbar)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5150 to 1.5170 (20°C, 589nm)
Linear Formula C2H5CH(C6H5)CO2H
Quantity 5 g
Beilstein 09,IV,1815
Specific Gravity 1.055
Solubility Information Solubility in water: insoluble
Formula Weight 164.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (R)-(-)-2-Phenylbutyric acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.