Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-3-Aminopiperidine Dihydrochloride 97.0+%, TCI America™
SDP

Catalog No. a2787200mg Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 334618-23-4
Molecular Formula C5H14Cl2N2
Molecular Weight (g/mol) 173.081
MDL Number MFCD06799458
InChI Key GGPNYXIOFZLNKW-ZJIMSODOSA-N
Synonym r-3-aminopiperidine dihydrochloride, r-piperidin-3-amine dihydrochloride, r---3-aminopiperidine dihydrochloride, r-3-piperidinamine dihydrochloride, 3r-piperidin-3-amine dihydrochloride, r-3-amino-piperidine dihydrochloride, r---3-aminopiperidine 2hcl, r-3-aminopiperidine 2hcl, r-3-amino piperidine dihcl
PubChem CID 16211333
IUPAC Name (3R)-piperidin-3-amine;dihydrochloride
SMILES C1CC(CNC1)N.Cl.Cl

Specifications

Melting Point 208°C
Color White-Yellow
Quantity 200 mg
Formula Weight 173.08
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-3-Aminopiperidine Dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.