Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine, 97%, Thermo Scientific Chemicals

Catalog No. AAH63889MD
Click to view available options
Quantity:
250 mg
5 g

Thiazoles are used in peptide research. Mainly used in medicine field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazoles are used in peptide research. Mainly used in medicine field.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. 2-7°C Refrigerate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1212136-00-9
Molecular Formula C19H21NO7S
Molecular Weight (g/mol) 407.437
MDL Number MFCD09864994
InChI Key RTHSATBSXDKZBN-QGZVFWFLSA-N
Synonym methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate, methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate
PubChem CID 46172874
IUPAC Name methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC

Specifications

Melting Point 100°C to 107°C
Color White to Brown
Quantity 250 mg
Formula Weight 237.27
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.