Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 98%
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an active pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as an active pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 170°C to 172°C |
| Quantity | 1 g |
| Solubility Information | Slightly soluble in water. |
| Optical Rotation | −182° (c=1 in DMF) |
| Formula Weight | 432.43 |
| Percent Purity | 98% |
| Chemical Name or Material | (R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction