Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-(Fmoc-amino)-3-phenylpropionic acid, 95%

Catalog No. p-7034391
Click to view available options
Quantity:
250 mg
1 g

CAS: 220498-02-2 | C24H21NO4 | 387.435 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220498-02-2
Molecular Formula C24H21NO4
Molecular Weight (g/mol) 387.435
MDL Number MFCD01311783
InChI Key PTSLRPMRTOVHAB-JOCHJYFZSA-N
Synonym r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-3-amino-3-phenylpropionic acid, fmoc-l-beta-phenylalanine, fmoc-beta-phe-oh, r-n-fmoc-3-amino-3-phenylpropanoic acid, r-3-fmoc-amino-3-phenylpropionic acid, fmoc-r-phenylalanine, 3r-3-9h-fluoren-9-ylmethoxycarbonylamino-3-phenylpropanoic acid, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid, fmoc-d-?-phe-oh
PubChem CID 2734302
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 250 mg
Beilstein 7982022
Formula Weight 387.43
Percent Purity 95%
Chemical Name or Material (R)-3-(Fmoc-amino)-3-phenylpropionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.