Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-4-Benzyloxycarbonylamino-2-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals

Catalog No. AAH6281703
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6281703 1 g
AAH6281706 5 g
AAH62817MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6281703 Supplier Thermo Scientific Chemicals Supplier No. H6281703

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101854-42-6
Molecular Formula C29H47N3O6
Molecular Weight (g/mol) 533.71
MDL Number MFCD00798628
InChI Key CBSVEVKFQHTZSP-BTQNPOSSSA-N
Synonym boc-d-dab z-oh.dcha, dicyclohexylamine r-4-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino butanoate, boc-d-dab z-oh*dcha, n-boc-n'-cbz-d-2,4-diaminobutyric acid dicyclohexylamine salt, r-2-1,1-dimethylethoxy carbonyl amino-4-phenylmethoxy carbonyl amino butanoic acid, dicyclohexylamine, n-a-boc-n-?-z-d-2,4-diaminobutyric aciddicyclohexylaminesalt, 2r-4-benzyloxycarbonylamino-2-tert-butoxycarbonylamino butanoic acid; n-cyclohexylcyclohexanamine, 2r-4-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2r-4-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino butanoic acid; dicha
PubChem CID 45357174
IUPAC Name N-cyclohexylcyclohexanamine;(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-(phenylmethoxycarbonylamino)butanoic acid
SMILES CC(C)(C)OC(=O)NC(CCNC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

Quantity 1 g
Formula Weight 533.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (R)-4-Benzyloxycarbonylamino-2-(Boc-amino)butyric acid dicyclohexylammonium salt
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.