Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ R 568 hydrochloride
GREENER_CHOICE

Catalog No. p-4816859
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-151-0 10 mg
38-155-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-151-0 Supplier Tocris Bioscience™ Supplier No. 3815/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Positive allosteric modulator of human calcium-sensing receptor (CaSR)

Positive allosteric modulator and allosteric agonist of the human calcium-sensing receptor (hCaSR). Exhibits calcimimetic activity. Has the ability to restore function to hCaSR mutants that cause familial hypocalciuric hypercalcemia (FHH) and neonatal severe hyperparathyroidism (NSHPT). Exerts a suppressive effect on parathyroid (PT) hormone secretion; inhibits PT cell proliferation in rats with renal insufficiency.

Chemical Identifiers

CAS 177172-49-5
Molecular Formula C18H23Cl2NO

Specifications

Quantity 10 mg
Formula Weight 340.29
Purity >99%
Molecular Weight (g/mol) 340.288
Chemical Name or Material R 568 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.