Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-6-Chloro-2-hexanol 98+% , Thermo Scientific™

Catalog No. AC390495000
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle

Chemical Identifiers

CAS 154885-33-3
Molecular Formula C6H13ClO
Molecular Weight (g/mol) 136.619
MDL Number MFCD04040052
InChI Key LDIPECSHAACCTQ-ZCFIWIBFSA-N
Synonym r-6-chloro-2-hexanol, 2r-6-chlorohexan-2-ol, r-6-chlorohexan-2-ol, r-1-chloro-5-hydroxyhexane, 2-hexanol,6-chloro-, r-9ci, 2-hexanol, 6-chloro-, r-9ci
PubChem CID 7016437
IUPAC Name (2R)-6-chlorohexan-2-ol
SMILES CC(CCCCCl)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Refractive Index 1.4510 to 1.4530
Linear Formula CH3CH(OH)(CH2)4Cl
Quantity 500 mg
Formula Weight 136.62
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material (R)-6-Chloro-2-hexanol
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.