Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (R)-(-)-Apomorphine hydrochloride
GREENER_CHOICESDP

Catalog No. 207350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-735-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 20-735-0 Supplier Tocris Bioscience™ Supplier No. 2073/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dopamine agonist; non-subtype-selective

Prototypical dopamine agonist (pKi values are 6.43, 7.08, 7.59, 8.36 and 7.83 for human recombinant D1, D2L, D3, D4 and D5 receptors respectively). Produces biphasic effects on locomotor activity, and displays anti-Parkinsonian and neuroprotective actions following systemic administration in vivo.

Chemical Identifiers

CAS 314-19-2
Molecular Formula C17H18ClNO2

Specifications

Quantity 50 mg
Formula Weight 303.79
Purity >99%
Molecular Weight (g/mol) 303.786
Chemical Name or Material (R)-(-)-Apomorphine hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.