Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-BINAP, TCI America™
SDP

Catalog No. p-7193340 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Specifications

CAS 76189-55-4
Melting Point 242°C
Color White-Yellow
Molecular Formula C44H32P2
MDL Number MFCD00010805
Quantity 5 g
Synonym binap, r-+-2,2 '-bis diphenylphosphino-1,1 '-binaphthyl, 2,2 '-bis diphenylphosphino-1,1 '-binaphthyl, s-binap, s---binap
InChI Key MUALRAIOVNYAIW-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8
IUPAC Name [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
Molecular Weight (g/mol) 622.688
PubChem CID 634876
Formula Weight 622.69
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.