Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (R)-Citalopram oxalate
GREENER_CHOICESDP

Catalog No. 576350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-635-0 50 mg
57-631-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-635-0 Supplier Tocris Bioscience™ Supplier No. 5763/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enantiomer of escitalopram oxalate (Cat. No. 1427)

(R)-Citalopram oxalate is an enantiomer of escitalopram oxalate . Postulated to allosterically modulate the affinity of the S-enantiomer.

Racemate and S-enantiomer also available.

Specifications

Chemical Name or Material (-)-(R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate
CAS 219861-53-7
Quantity 50 mg
Target 5-HT Transporter Inhibitors
Molecular Formula C20H21FN2O.C2H2O4
Formula Weight 414.43
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.